Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-ITPR3 Rabbit Polyclonal Antibody
  BSBTPB9428
undefined
Anti-ITPR3 Rabbit Polyclonal Antibody
  BSBTPB9428

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    inositol 1,4,5-trisphosphate receptor, type 3
  • Symbol antigenu:
    ITPR3
  • Klonovalita:
    Polyclonal
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Formulář:
    Lyophilized
  • Identifikační číslo genu:
    3710
  • Synonyma antigenu:
    IP3R3|IP3R
  • UniProtKB:
    Q14573
  • Skladovací teplota:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Imunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human ITPR3 (70-95aa QYWKAKQTKQDKEKIADVVLLQKLQH), identical to the related mouse and rat sequences.
  • Čištění:
    Immunogen affinity purified.
  • Rozměry protilátky:
    100 µg/vial
  • Bal.:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Inositol 1,4,5-trisphosphate receptor type 3(ITPR3) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

ITPR3 encodes a receptor for inositol 1,4,5-trisphosphate, a second messenger that mediates the release of intracellular calcium. The receptor contains a calcium channel at the C-terminus and the ligand-binding site at the N-terminus. Knockout studies in mice suggest that type 2 and type 3 inositol 1,4,5-trisphosphate receptors play a key role in exocrine secretion underlying energy metabolism and growth. ITP3 channels serve an important role in the taste transduction pathway of sweet, bitter and umami tastes the gustatory system. ITP3 channels allow the flow of Calcium out of the endoplasmic reticulum in response to IP3. Calcium cations result in the activation of TRPM5 which leads to a depolarisation generating potential and an action potential.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for ITPR3 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: KDR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human