- Druh protilátky:Primární
- Název antigenu:major histocompatibility complex, class I, A
- Symbol antigenu:HLA-A
- Klonovalita:Polyclonal
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Formulář:Lyophilized
- Identifikační číslo genu:3105
- Synonyma antigenu:HLAA
- UniProtKB:P01892
- Skladovací teplota:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Imunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human HLA A (82-106aa EYWDGETRKVKAHSQTHRVDLGTLR).
- Čištění:Immunogen affinity purified.
- Rozměry protilátky:100 µg/vial
- Bal.:100 µG
Rabbit IgG polyclonal antibody for HLA class I histocompatibility antigen, A-2 alpha chain(HLA-A) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
HLA-A, namely HLA A, is a group of human leukocyte antigens (HLA) that are coded for by the HLA-A locus, which is located at human chromosome 6p21.3. HLA is simply the major histocompatibility complex (MHC) specific to humans. HLA-A is one of three major types of human MHC class I cell surface receptors. The others are HLA-B and HLA-C. And it is critical to the cytotoxic t-cell controlled immune response to viruses and other intracellular pathogens. Because each HLA-A gene has a high affinity for slightly different peptides, certain HLA-As are associated with increased risk, more rapid progression, and/or increased severity of many diseases. For similar reasons, HLA-A matching is essential to successful tissue transplants.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
WB: The detection limit for HLA A is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: HLA-C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat