Specifications
- Druh protilátky:Primární
- Název antigenu:Integrin Alpha 1
- Symbol antigenu:ITGA1
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Epitop:C-Terminal
- Křížová adsorbce:No
- Formulář:Lyophilized
- Synonyma antigenu:VLA1|CD49a
- Počet aminokyselin:1121 to 1134
- UniProtKB:P56199
- Sekvence:SNQKRELAIQISKD
- Skladovací teplota:–20 °C
- Imunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Integrin alpha 1 (1121-1134aa SNQKRELAIQISKD), different from the related mouse and rat sequences by one amino acid.
- Čištění:purified by immunogen affinity purification
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Integrin alpha-1(ITGA1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Integrin alpha 1(ITGA1) chain associates with the beta 1(ITGB1) chain to form a heterodimer that functions as a dual laminin/collagen receptor in neural cells and hematopoietic cells. ITGA1 has a 206-amino acid I domain in its N-terminal half, followed by 3 divalent cation-binding sites and a C-terminal transmembrane domain with a short cytoplasmic tail. It also has 28 potential N-glycosylation sites. Human ITGA1 was expressed in a mouse fibroblast cell line as a 180-kD protein. ITGA1 is involved in the early remodeling of osteoarthritic cartilage and plays an essential role in the regulation of mesenchymal stem cell proliferation and cartilage production. It also plays an essential role in the regulation of MSC proliferation and cartilage production.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: Integrin alpha 1
Clonality:
Clone:
Conjugation:
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat