ContactUsLinkComponent
Stoppers, butyl, for diagnostic vials, WHEATON®
Stoppers, butyl, for diagnostic vials, WHEATON®
Catalog # LSUK600005
Supplier:  DWK Life Sciences
undefined
Stoppers, butyl, for diagnostic vials, WHEATON®
Catalog # LSUK600005
Supplier:  DWK Life Sciences
Specifications
  • Ø horní části:
    14 mm
  • Popis:
    Freeze dry stopper
  • Sekvence:
    01
  • Bal.:
    1.000
Specifications
About this item
Stoppers are made from the pharmaceutical grade materials.
  • Suitable for freeze drying