Wipes for general applications, Polytex® light
Catalog # ZVGA10160-00
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Specifications
- Balené:Pack of 420 (12×35 wipes)
- Barva:Bílá
- D׊:400×420 mm
- Bal.:420
Specifications
About this item
These solvent resistant wipes made of 100% PP are suitable for wet wiping in a variety of applications.
- Durable and lint-free
- Even remove stubborn dirt
- Suitable for food contact
Ideal for use in garages, paint shops, catering areas, kitchens and food processing areas.