Temperature indicating kit
Thermax irreversible research kit
Katalogové číslo 620-9129
Dodavatel: KLEINFELD
Přejít na:
Product Details & Documents
A compilation of all thermal colors and a pattern of stripes for eight temperatures in each range.
: Content includes 13×15 ml vials with either colours with single envelope or multiple envelope, 15 ml diluent, a brush for applying and a lab-pack with 25 self-adhesive indicators.
Specifikace
- Popis:Thermax irreversible research kit