Culture media in bottles, CHROMagar™
CHRASB283-25
New Product
:
- Bal.:25 L
- Popis:CHROMagar™ Strep B
- Sekvence:4
- Kat. č.:SB283-25
- Náplň:25 L
With traditional media identification of mixed cultures is difficult, whereas CHROMagar™ just by the colour of the colony will instantly differentiate various species.
- High contrast
- Fast
- Convenient