Orders LinkContactUsLinkComponent
Tweezers, reverse action
Tweezers, reverse action
Straight, round tips, 321R, 125 mm
Catalog # OUTI321R
Supplier:   OUTILS RUBIS
Tweezers, reverse action
Catalog # OUTI321R
Supplier:  OUTILS RUBIS
Tweezers, reverse action
Tweezers, reverse action
Straight, round tips, 321R, 125 mm
Price
766,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Comfortable soldering tweezers with reversed action.
Specifications
  • Type:
    321R
  • Popis:
    Straight, round tips
  • Délka:
    125 mm