Orders LinkContactUsLinkComponent
pH combination electrodes, IntelliCAL™, High Performance, Red Rod
Dodavatel:   Hach
pH combination electrodes, IntelliCAL™, High Performance, Red Rod
Dodavatel:  Hach
Certifikáty
pH combination electrodes, IntelliCAL™, High Performance, Red Rod
pH combination electrodes, IntelliCAL™, High Performance, Red Rod
7 Dostupných variant níže
Product Details & Documents
Digital high-performance pH electrodes with Red Rod reference system and built-in temperature sensor.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Red Rod system creates a trouble-free liquid junction
  • Encapsulation of reference elements: no silver ions and therefore less risk of clogging when measuring in challenging sample matrices
  • Electrodes store time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
  • Electrodes can easily be moved between different HQD meters
  • Ideal for a multi-user environment with multiple HQD series laboratory and portable meters
The symmetry formed by the internal and external Red Rod reference elements means that the iso pH is the same as the zero pH which provides highly reproducible results and a fast, accurate response, even when temperature fluctuations occur. Red Rod electrodes provide a stable signal by protecting the reference elements from light interference. The saturated KCl salt-bridge also helps in creating a stable liquid junction potential.
Product Family Options
Informace o produktu
  • Sekvence
    Type
    pH
    Tepl.
    Materiál
    Elektrolyt
    Diafragma
    Ø×D
    Připojení
    Dostupnost
    Price
  • 665-0384
    pH combination electrode, IntelliCAL™ PHC705
    pH combination electrode, IntelliCAL™ PHC705
    20
    IntelliCAL™ PHC705, Red Rod
    0 - 14
    −10...+100 °C
    Sklo
    Sat. KCl (refilable)
    Porous pin
    12×200 mm
    1 m cable
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 150,00 Kč
    20 150,00 Kč
    1 * 1 KS
       20 150,00 Kč
  • 665-0385
    pH combination electrode, IntelliCAL™ PHC705A
    pH combination electrode, IntelliCAL™ PHC705A
    21
    IntelliCAL™ PHC705A, Red Rod
    0 - 14
    −10...+100 °C
    Sklo
    Sat. KCl (refilable)
    Porous pin
    12×200 mm
    1 m cable
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 150,00 Kč
    20 150,00 Kč
    1 * 1 KS
       20 150,00 Kč
  • 665-0386
    pH combination electrode, IntelliCAL™ PHC725
    pH combination electrode, IntelliCAL™ PHC725
    22
    IntelliCAL™ PHC725, Red Rod
    0 - 14
    −10...+100 °C
    Sklo
    Sat. KCl (refilable)
    Annular ring
    12×200 mm
    1 m cable
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 150,00 Kč
    20 150,00 Kč
    1 * 1 KS
       20 150,00 Kč
  • 665-0387
    pH combination electrode, IntelliCAL™ PHC729
    pH combination electrode, IntelliCAL™ PHC729
    23
    IntelliCAL™ PHC729, Red Rod
    0 - 12
    0...+100 °C
    Sklo
    Sat. KCl (refilable)
    Annular ring
    12×200 mm
    1 m cable
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 20 150,00 Kč
    20 150,00 Kč
    1 * 1 KS
       20 150,00 Kč
  • 665-0388
    pH combination electrode, IntelliCAL™ PHC735
    pH combination electrode, IntelliCAL™ PHC735
    24
    IntelliCAL™ PHC735, Red Rod
    0 - 14
    −10...+100 °C
    Sklo
    Sat. KCl (refilable)
    Annular ring
    12×200 mm
    1 m cable
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 22 910,00 Kč
    22 910,00 Kč
    1 * 1 KS
       22 910,00 Kč
  • 665-0389
    pH combination electrode, IntelliCAL™ PHC745
    pH combination electrode, IntelliCAL™ PHC745
    25
    IntelliCAL™ PHC745, Red Rod
    0 - 14
    −10...+100 °C
    Sklo
    Sat. KCl (refilable)
    Sleeve
    12×200 mm
    1 m cable
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 22 600,00 Kč
    22 600,00 Kč
    1 * 1 KS
       22 600,00 Kč
  • 665-0390
    pH combination electrode, IntelliCAL™ PHC805
    pH combination electrode, IntelliCAL™ PHC805
    26
    IntelliCAL™ PHC805, Red Rod
    0 - 14
    0...+80 °C
    Sklo
    3M KCl (refilable)
    Porous pin
    12×200 mm
    1 m cable
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 13 060,00 Kč
    13 060,00 Kč
    1 * 1 KS
       13 060,00 Kč
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Bal.
    Dostupnost
    Price
  • Roztok nasycený, KCL, 30 ml, láhev
    Roztok nasycený, KCL, 30 ml, láhev
    HACHS21M010
    02
    30 mL
    Přihlásit se pro dostupnost
    1 * 30 mL - 565,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Bal.
    Dostupnost
    Price
  • Roztok KCL 3M, nasycený AgCl, pro Ag/AgCl referenční elementy elektrody (pHC3xxx/REF3xx), láhev 100 ml
    Roztok KCL 3M, nasycený AgCl, pro Ag/AgCl referenční elementy elektrody (pHC3xxx/REF3xx), láhev 100 ml
    HACHS21M004
    05
    100 mL
    Přihlásit se pro dostupnost
    1 * 100 mL - 803,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Bal.
    Dostupnost
    Price
  • Extra strong electrode cleaning solution, RENOVO.X, 250 ml bottle
    Extra strong electrode cleaning solution, RENOVO.X, 250 ml bottle
    HACHS16M002
    99
    250 mL
    Přihlásit se pro dostupnost
    1 * 250 mL - 803,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Bal.
    Dostupnost
    Price
  • Mild cleaning solution for electrodes, RENOVO.N, 250 ml bottle
    Mild cleaning solution for electrodes, RENOVO.N, 250 ml bottle
    HACHS16M001
    98
    250 mL
    Přihlásit se pro dostupnost
    1 * 250 mL - 803,00 Kč
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • KCl solution, saturated, 59 ml bottle
    KCl solution, saturated, 59 ml bottle
    HACH25118026
    Přihlásit se pro dostupnost
    1 * 1 KIT