Supelco® Ascentis® Express C8 HPLC and UHPLC Columns
: Merck
Ascentis® Express C8 high-speed, high-performance liquid chromatography columns are based on the highly efficient Fused-Core® particle design.
The Fused-core particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0,6 µm thick porous shell and the highly uniform overall particle size.
The densely bonded, extensively endcapped dimethyloctyl stationary phase of Ascentis Express 5 μm C8 provides a stable, reversed-phase packing that can be used for basic, acidic or neutral compounds.
: Supplied with column test certificate inside the box.
- Catalog No:
- 50396-U
- 50362-U
- 53857-U
- 50394-U
- 50395-U
- 53983-U
- 53831-U
- 53852-U
- 50392-U
- 53853-U
- 53848-U
- 53849-U
- 54272-U
- 50381-U
- 53511-U
- 50382-U
- 53509-U
- 50367-U
- 53837-U
- 50368-U
- 53838-U
- 50363-U
- 53834-U
- 50364-U
- 53836-U
- 50377-U
- 53844-U
- 50378-U
- 53839-U
- 53843-U
- 50376-U
- 54260-U
- 50399-U
- 53512-U
- 53832-U
- 51657-U
- 53858-U
- 51654-U
- 51656-U
- 50390-U
- 50391-U
- 50386-U
- 50389-U
- 51652-U
- 50385-U
- 53795-U
- 51679-U
- 51663-U
- 51664-U
- 51658-U
- 51661-U
- 51674-U
- 51676-U
- 51672-U
- 53997-U
- 51673-U
- 50372-U
- Popis:
- 5,0 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 2,7 µm columns
- 2,0 µm columns
- 2,7 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 5,0 µm columns
- 2,0 µm columns
- 5,0 µm columns
- 2,7 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,0 µm columns
- 2,7 µm columns
- 2,0 µm columns
- 5,0 µm columns
- Naplnění uhlíkem:
- 3,7%
- 3,7%
- 5,4%
- 3,7%
- 3,7%
- 5,4%
- 5,4%
- 5,4%
- 3,7%
- 5,4%
- 5,4%
- 5,4%
- 5,4%
- 3,7%
- 5,4%
- 3,7%
- 5,4%
- 3,7%
- 5,4%
- 3,7%
- 5,4%
- 3,7%
- 5,4%
- 3,7%
- 5,4%
- 3,7%
- 5,4%
- 3,7%
- 5,4%
- 5,4%
- 3,7%
- 5,4%
- 3,7%
- 5,4%
- 5,4%
- 4,8%
- 5,4%
- 4,8%
- 4,8%
- 3,7%
- 3,7%
- 3,7%
- 3,7%
- 4,8%
- 3,7%
- 5,4%
- 4,8%
- 4,8%
- 4,8%
- 4,8%
- 4,8%
- 4,8%
- 4,8%
- 4,8%
- 5,4%
- 4,8%
- 3,7%
- S navazanou skupinou:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Maximální tlak:
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 1000 bar
- 600 bar
- 1000 bar
- 1000 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 1000 bar
- 600 bar
- 600 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 600 bar
- 1000 bar
- 600 bar
- Rozsah pH:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Povrchová oblast:
- 90 m²/g
- 90 m²/g
- 135 m²/g
- 90 m²/g
- 90 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 90 m²/g
- 135 m²/g
- 135 m²/g
- 120 m²/g
- 135 m²/g
- 120 m²/g
- 120 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 90 m²/g
- 120 m²/g
- 90 m²/g
- 135 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 120 m²/g
- 135 m²/g
- 120 m²/g
- 90 m²/g
- Kód USP:
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7