Microorganisms strains, Culti-Loops®
OXOICL7050
:
- Bal.:5
- Popis:Escherichia coli ATCC® 25922™*
Culti-Loops® are ready-to-use, disposable inoculation loops containing stabilized, preserved, viable microorganisms. A quality source of stock organisms, Culti-Loops® are derived from American Type Culture Collection (ATCC®) or other referenced sources. Culti-Loops® are manufactured in an ISO-certified facility using our proprietary preservation technique, ensuring enhanced recovery and reproducibility. The loop may be dissolved in rehydration fluid or streaked directly onto appropriate media.
- Convenient, ready-to-use, disposable loops
- Organisms include bacterial, fungal, and mycobacterium isolates
- Individually wrapped in foil-sealed pouches to preserve viability
- Peel-off label for convenient documentation
- No glass, needles, or forceps ensures user safety