Order Entry
Czech Republic
ContactUsLinkComponent
Anti-N-myc Downstream Regulated Gene 1 Protein Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anti-N-myc Downstream Regulated Gene 1 Protein Rabbit Polyclonal Antibody (PE (Phycoerythrin))
  USBIN2915-52N-PE
undefined
Anti-N-myc Downstream Regulated Gene 1 Protein Rabbit Polyclonal Antibody (PE (Phycoerythrin))
  USBIN2915-52N-PE

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    N-myc Downstream Regulated Gene 1 Protein
  • Klonovalita:
    Polyclonal
  • Spojení:
    PE (Phycoerythrin)
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Epitop:
    N-terminal
  • Formulář:
    liquid
  • Skladovací pufr:
    PBS, pH 7,2
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Testované aplikace:
    Fluoresence ELISA
  • Čištění:
    purified by immunoaffinity chromatography
  • Rozměry protilátky:
    200 µl
  • Bal.:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: N-myc Downstream Regulated Gene 1 Protein
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human