Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MHC Class 1 Chain-related Gene A Rabbit Polyclonal Antibody
Anti-MHC Class 1 Chain-related Gene A Rabbit Polyclonal Antibody
Catalog # USBIM3886-75A
Supplier:  US Biological
undefined
Anti-MHC Class 1 Chain-related Gene A Rabbit Polyclonal Antibody
Catalog # USBIM3886-75A
Supplier:  US Biological

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    MHC Class 1 Chain-related Gene A
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Rabbit
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Formulář:
    liquid
  • Počet aminokyselin:
    68 to 97
  • Skladovací pufr:
    PBS, 0,09% sodium azide.
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Čištění:
    purified by Protein A chromatography
  • Rozměry protilátky:
    200 µl
  • Bal.:
    200 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-1:50; Flow Cytometry: 1:10-1:50; Immunofluorescence: 1:10-1:50

Type: Primary
Antigen: MHC Class 1 Chain-related Gene A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human