Anti-MEK1, MEK2 Rabbit Polyclonal Antibody
Catalog # USBIM2865-10G
Supplier: US Biological
Specifications
- Druh protilátky:Primární
- Symbol antigenu:MEK1/2
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Formulář:liquid
- Modifikace:phospho-Ser222
- Skladovací pufr:PBS, pH 7,2, 1mg/ml BSA (IgG free, proteasefree), 0,05% sodium azide.
- Skladovací teplota:Cool
- Koncentrace:~0,05 mg/ml
- Transportní teplota:Ice
- Čištění:purified by immunoaffinity chromatography
- Rozměry protilátky:10 Pieces
- Bal.:10
Specifications
About this item
Recommended dilution: Western Blot: 0.1-1 ?g/ml
Type: Primary
Antigen: MEK1, MEK2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat