Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SMEK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-SMEK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI042042-FITC
Supplier:  US Biological
Anti-SMEK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI042042-FITC
Supplier:  US Biological

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    SMEK homolog 2, suppressor of mek1 (Dictyostelium)
  • Symbol antigenu:
    SMEK2
  • Klonovalita:
    Polyclonal
  • Spojení:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Formulář:
    liquid
  • Počet aminokyselin:
    719 to 748
  • Skladovací pufr:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Testované aplikace:
    Fluoresence ELISA
  • Čištění:
    purified by Protein A chromatography
  • Rozměry protilátky:
    200 µl
  • Bal.:
    200 µl

Specifications

About this item

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: SMEK2 (SMEK homolog 2, suppressor of mek1 (Dictyostelium))
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human