Anti-AKT3 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 25F6.F6.D8]
Catalog # ROCK200-302-E75
Supplier: Rockland Immunochemicals
Specifications
- Druh protilátky:Primární
- Název antigenu:v-akt murine thymoma viral oncogene homolog 3
- Symbol antigenu:AKT3
- Klonovalita:Monoclonal
- Klon:25F6.F6.D8
- Spojení:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Průtoková cytometrie:Yes
- Hostitel:Mouse
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Izotyp:IgG1 kappa
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Epitop:Internal
- Křížová adsorbce:none
- Formát:Antibody fraction
- Formulář:Lyophilized
- Identifikační číslo genu:10000
- Synonyma antigenu:PKB-GAMMA|PKBG|RAC-PK-gamma|STK-2|RAC-gamma|PRKBG|MPPH2|MPPH
- Skladovací pufr:0,02 M Potassium Phosphate, 0,5 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Skladovací teplota:Cool
- Koncentrace:1,0 mg/ml by UV absorbance at 280 nm
- Transportní teplota:Ice
- Imunogen:Anti-AKT3 Antibody was prepared from tissue culture supernatant by Protein A affinity chromatography using a synthetic peptide corresponding to internal residues of human AKT3 Protein.
- Testované aplikace:Fluoresence ELISA
- Čištění:Protein A purified
- Rozměry protilátky:50 µg
- Bal.:50 µG
Specifications
About this item
Anti-AKT3 FITC Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT3 Protein by western blotting in the appropriate cell lysate or extract.
Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl to 20 µl
Type: Primary
Antigen: AKT3
Clonality: Monoclonal
Clone: 25F6.F6.D8
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Center
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse, Rat, Human