Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IgG F(c) Goat Polyclonal Antibody
Anti-IgG F(c) Goat Polyclonal Antibody
  ROCK712-1122
undefined
Anti-IgG F(c) Goat Polyclonal Antibody
  ROCK712-1122

 

  • Druh protilátky:
    Sekundární
  • Symbol antigenu:
    IgG F(c)
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Průtoková cytometrie:
    Yes
  • Posun gelu:
    Yes
  • Hostitel:
    Goat
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Imunoprecipitace:
    Yes
  • Reaktivita:
    Rat
  • Western blot:
    Yes
  • Křížová adsorbce:
    Bovine, Horse, Human
  • ChIP:
    Yes
  • Neutralizace:
    Yes
  • Formát:
    Antibody fraction
  • Formulář:
    Liquid (sterile filtered)
  • Fragment protilátky:
    F(ab')2
  • Skladovací pufr:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Skladovací teplota:
    Cool
  • Koncentrace:
    1,0 mg/ml by UV absorbance at 280 nm
  • Transportní teplota:
    Ice
  • Imunogen:
    Anti-Rat IgG F(c) was produced by repeated immunization with Rat F(c) fragment in goat.
  • Čištění:
    purified by immunoaffinity chromatography
  • Rozměry protilátky:
    1 mg
  • Bal.:
    1 mg

 

 

Rat Fc secondary antibody is suitable for immunoblotting, western blot, dot blot, ELISA, and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:5000; Immunochemistry: 1:1000 - 1:3000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rat