Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Katalogové číslo ROCK711-1622
undefined
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Katalogové číslo ROCK711-1622

Specifikace

  • Druh protilátky:
    Sekundární
  • Symbol antigenu:
    IgG (H+L)
  • Klonovalita:
    Polyclonal
  • Spojení:
    Biotin
  • ELISA:
    Yes
  • Průtoková cytometrie:
    Yes
  • Posun gelu:
    Yes
  • Hostitel:
    Goat
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Imunoprecipitace:
    Yes
  • Reaktivita:
    Rabbit
  • Western blot:
    Yes
  • ChIP:
    Yes
  • Neutralizace:
    Yes
  • Formát:
    Antibody fraction
  • Formulář:
    Lyophilized
  • Fragment protilátky:
    F(ab')2
  • Skladovací pufr:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Skladovací teplota:
    Cool
  • Koncentrace:
    1,0 mg/ml by UV absorbance at 280 nm
  • Transportní teplota:
    Ice
  • Imunogen:
    Anti-Rabbit IgG F(ab')2 fragment was produced by repeated immunization with Rabbit IgG whole molecule in goat.
  • Čištění:
    purified by immunoaffinity chromatography
  • Rozměry protilátky:
    500 µg
  • Bal.:
    500 µG

Specifikace

O tomto produktu

F(ab')2 anti-Rabbit IgG is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays. Specific conditions for reactivity and signal detection should be optimized by the end user.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit