Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-kappa light chain Goat Polyclonal Antibody
  ROCK709-1110
Anti-kappa light chain Goat Polyclonal Antibody
  ROCK709-1110

 

  • Druh protilátky:
    Sekundární
  • Název antigenu:
    Kappa Light Chain
  • Symbol antigenu:
    IGKC
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Průtoková cytometrie:
    Yes
  • Posun gelu:
    Yes
  • Hostitel:
    Goat
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Imunoprecipitace:
    Yes
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Křížová adsorbce:
    none
  • ChIP:
    Yes
  • Neutralizace:
    Yes
  • Formát:
    Antibody fraction
  • Formulář:
    Liquid (sterile filtered)
  • Fragment protilátky:
    F(ab')2
  • Skladovací pufr:
    0,125 M Sodium Borate, 0,075 M Sodium Chloride, 0,005 M EDTA, pH 8.0, 0,01% (w/v) Sodium Azide,
  • Skladovací teplota:
    Cool
  • Koncentrace:
    1,0 mg/ml by UV absorbance at 280 nm
  • Transportní teplota:
    Ice
  • Imunogen:
    Anti-Human IgG (kappa chain) was produced by repeated immunization with human IgG kappa light chain in goat.
  • Čištění:
    purified by immunoaffinity chromatography
  • Rozměry protilátky:
    1 mg
  • Bal.:
    1 mg

 

 

Antibody Anti-Human IgG (kappa chain) is suitable for immunoblotting (western or dot blot), ELISA, and immunochemistry assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent);

Type: Secondary
Antigen: Igκ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Light Chain
Host: Goat
Isotype:
Reactivity: Human