Order Entry
Czech Republic
ContactUsLinkComponent
Anti-FITC Goat Polyclonal Antibody (Texas Red®)
Anti-FITC Goat Polyclonal Antibody (Texas Red®)
  ROCK700-109-096
Anti-FITC Goat Polyclonal Antibody (Texas Red®)
  ROCK700-109-096

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Fluorescein
  • Symbol antigenu:
    FITC
  • Klonovalita:
    Polyclonal
  • Spojení:
    Texas Red®
  • ELISA:
    Yes
  • Hostitel:
    Goat
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Reaktivita:
    Multiple
  • Western blot:
    Yes
  • Křížová adsorbce:
    none
  • Formát:
    Antibody fraction
  • Formulář:
    Lyophilized
  • Fragment protilátky:
    F(ab')2
  • Skladovací pufr:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Skladovací teplota:
    Cool
  • Koncentrace:
    1,0 mg/ml by UV absorbance at 280 nm
  • Transportní teplota:
    Ice
  • Imunogen:
    Fluorescein conjugated to Goat IgG
  • Čištění:
    purified by immunoaffinity chromatography
  • Rozměry protilátky:
    1 mg
  • Bal.:
    1 mg

 

 

Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: 1:500 - 1:2500; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: FITC
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: pan