Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IgG F(c) Rabbit Polyclonal Antibody (Texas Red®)
Anti-IgG F(c) Rabbit Polyclonal Antibody (Texas Red®)
  ROCK614-4903
Anti-IgG F(c) Rabbit Polyclonal Antibody (Texas Red®)
  ROCK614-4903

 

  • Druh protilátky:
    Sekundární
  • Symbol antigenu:
    IgG F(c)
  • Klonovalita:
    Polyclonal
  • Spojení:
    Texas Red®
  • ELISA:
    Yes
  • Průtoková cytometrie:
    Yes
  • Posun gelu:
    Yes
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Imunoprecipitace:
    Yes
  • Reaktivita:
    Pig
  • Western blot:
    Yes
  • Křížová adsorbce:
    none
  • ChIP:
    Yes
  • Neutralizace:
    Yes
  • Formát:
    Antibody fraction
  • Formulář:
    Lyophilized
  • Skladovací pufr:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Skladovací teplota:
    Cool
  • Koncentrace:
    2,0 mg/ml by UV absorbance at 280 nm
  • Transportní teplota:
    Ice
  • Imunogen:
    Swine IgG F(c) fragment
  • Čištění:
    purified by immunoaffinity chromatography
  • Rozměry protilátky:
    2 mg
  • Bal.:
    2 mg

 

 

Secondary Rabbit Anti-IgG F(c) Reacts with Pig (Swine, Porcine)

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Pig