Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Peroxidase)
  ROCK614-4304
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Peroxidase)
  ROCK614-4304

 

  • Druh protilátky:
    Sekundární
  • Symbol antigenu:
    IgG F(ab')2
  • Klonovalita:
    Polyclonal
  • Spojení:
    Peroxidase
  • ELISA:
    Yes
  • Průtoková cytometrie:
    Yes
  • Posun gelu:
    Yes
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Imunoprecipitace:
    Yes
  • Reaktivita:
    Pig
  • Western blot:
    Yes
  • Křížová adsorbce:
    none
  • ChIP:
    Yes
  • Neutralizace:
    Yes
  • Formát:
    Antibody fraction
  • Formulář:
    Lyophilized
  • Skladovací pufr:
    0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Skladovací teplota:
    Cool
  • Koncentrace:
    2,0 mg/ml by UV absorbance at 280 nm
  • Transportní teplota:
    Ice
  • Imunogen:
    Swine IgG F(ab')2 fragment
  • Čištění:
    purified by immunoaffinity chromatography
  • Rozměry protilátky:
    2 mg
  • Bal.:
    2 mg

 

 

This product has been assayed against 1.0 µg of Swine IgG F(ab')2  in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Pig