Order Entry
Czech Republic
ContactUsLinkComponent
Anti-AKR1C4 Goat Polyclonal Antibody
Anti-AKR1C4 Goat Polyclonal Antibody
Catalog # EVBIEB07240
Supplier:  Everest Biotech
undefined
Anti-AKR1C4 Goat Polyclonal Antibody
Catalog # EVBIEB07240
Supplier:  Everest Biotech

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    aldo-keto reductase family 1, member C4
  • Symbol antigenu:
    AKR1C4
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Goat
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Epitop:
    N-terminal
  • Křížová adsorbce:
    none
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    100 µg
  • Bal.:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: AKR1C4 (aldo-keto reductase family 1, member C4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human