Anti-GOPC Goat polyclonal antibody unconjugated
EVBIEB06239
- Druh protilátky:Primární
- Název antigenu:Golgi-associated PDZ and coiled-coil motif containing
- Symbol antigenu:GOPC
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Goat
- ImunoFluorescence:Yes
- Izotyp:IgG
- Western blot:Yes
- Epitop:C-terminal
- Křížová adsorbce:none
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:100 µg
- Bal.:0,1 mg
Type: Primary
Antigen: GOPC (golgi-associated PDZ and coiled-coil motif containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: