Anti-LAMP2 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: H4B4]
Catalog # NOVUNBP1-28553
Supplier: Novus Biologicals
Specifications
- Druh protilátky:Primární
- Název antigenu:Lysosomal-associated membrane protein 2
- Symbol antigenu:LAMP2
- Klonovalita:Monoclonal
- Klon:H4B4
- Spojení:FITC (Fluorescein Isothiocyanate)
- Průtoková cytometrie:Yes
- Hostitel:Mouse
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Izotyp:IgG1 kappa
- Reaktivita:Human
- Western blot:Yes
- Environmentally Preferable:
- Identifikační číslo genu:3920
- Synonyma antigenu:LAMP-2|LGP-B|Lysosome-associated membrane protein 2|CD107 antigen-like family member B|LAMP2|Lysosome-associated membrane glycoprotein 2|CD107b|Lysosomal membrane glycoprotein type B
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:1 mL
- Bal.:100 Tests
Specifications
About this item
Type: Primary
Antigen: LAMP2 (lysosomal-associated membrane protein 2)
Clonality: monoclonal
Clone: H4B4
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human