Anti-ABCE1 Rabbit Polyclonal Antibody (DyLight® 650)
NOVUNB400-116C
- Druh protilátky:Primární
- Název antigenu:ATP Binding Cassette, Subfamily E, Member 1
- Symbol antigenu:ABCE1
- Klonovalita:Polyclonal
- Spojení:DyLight® 650
- Hostitel:Rabbit
- Izotyp:
- Reaktivita:Human
- Western blot:Yes
- Environmentally Preferable:
- Identifikační číslo genu:6059
- Synonyma antigenu:RNase 4 inhibitor|Ribonuclease 4 Inhibitor|ABC 38|ABCE 1|OABP|ATP Binding Cassette Sub Family E Member 1|RLI|RNS4I|ABC38|RNase L Inhibitor
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:100 µL
- Bal.:0,1 mL
Type: Primary
Antigen: ABCE1 (ATP-binding cassette, sub-family E (OABP), member 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human