Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLC22A1 Rabbit Polyclonal Antibody
Anti-SLC22A1 Rabbit Polyclonal Antibody
Katalogové číslo BSBTPA2172
Dodavatel:  Boster Bio
undefined
Anti-SLC22A1 Rabbit Polyclonal Antibody
Katalogové číslo BSBTPA2172
Dodavatel:  Boster Bio

Specifikace

  • Druh protilátky:
    Primární
  • Název antigenu:
    Solute Carrier Family 22 Member A1
  • Symbol antigenu:
    Solute carrier family 22, member 1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-Terminal
  • Křížová adsorbce:
    No
  • Formulář:
    Lyophilized
  • Synonyma antigenu:
    HOCT1|SLC22A1|OCT1|oct1_cds
  • Počet aminokyselin:
    533 - 550
  • Sekvence:
    RKAKPKENTIYLKVQTSE
  • Skladovací teplota:
    –20 °C
  • Transportní teplota:
    Ice
  • Imunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human SLC22A1 (533-550aa RKAKPKENTIYLKVQTSE), different from the related mouse and rat sequences by six amino acids
  • Čištění:
    Purified by immunogen affinity purification
  • Rozměry protilátky:
    100 µg
  • Bal.:
    0,1 mg

Specifikace

O tomto produktu

Rabbit IgG polyclonal antibody for Solute carrier family 22 member 1(SLC22A1) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: SLC22A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human