Order Entry
Czech Republic
ContactUsLinkComponent
Anti-TIM 1 Rabbit Polyclonal Antibody
Anti-TIM 1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1624
Supplier:  Boster Bio
undefined
Anti-TIM 1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1624
Supplier:  Boster Bio

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Hepatitis A Virus Cellular Receptor 1
  • Symbol antigenu:
    TIM1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-Terminal
  • Křížová adsorbce:
    No
  • Formulář:
    Lyophilized
  • Synonyma antigenu:
    Tim-1
  • Počet aminokyselin:
    332 to 348
  • UniProtKB:
    Q96D42
  • Sekvence:
    IKALQNAVEKEVQAEDN
  • Skladovací teplota:
    –20 °C
  • Transportní teplota:
    Ice
  • Imunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human TIM 1 (332-348aa IKALQNAVEKEVQAEDN).
  • Čištění:
    purified by immunogen affinity purification
  • Rozměry protilátky:
    100 µg
  • Bal.:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Hepatitis A virus cellular receptor 1(HAVCR1) detection. Tested with WB, IHC-P, ICC in Human.

KIM1(KIDNEY INJURY MOLECULE 1), also known as HAVCR1, HAVCR or TIM1, is a protein that in humans is encoded by the KIM1 gene. The KIM1 gene is mapped to 5q33.3. Biochemical, mutational, and cell adhesion analyses confirm that Tim1 is capable of homophilic Tim-Tim interactions. The features identified in murine KIM1 is conserved in human KIM1. The KIM1 protein is indeed a receptor for the virus through the infection of canine osteogenic sarcoma cells expressing HAVCR1 with HAV. Using a monoclonal antibody to mouse Tim1, Tim1 is expressed after activation of naive T cells and on T cells differentiated in Th2-polarizing conditions. Ectopic expression of KIM1 during mouse T-cell differentiation leads to production of the Th2-type cytokine Il4, but not the Th1-type cytokine Ifng. KIM1-expressing epithelial cells internalized apoptotic bodies, and Kim1 is directly responsible for phagocytosis in cultured primary rat tubule epithelial cells and in porcine and canine epithelial cell lines.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: TIM 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human