Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-GDNF Rabbit Polyclonal Antibody
  BSBTPA1465
undefined
Anti-GDNF Rabbit Polyclonal Antibody
  BSBTPA1465

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Glial cell derived neurotrophic factor
  • Symbol antigenu:
    GDNF
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Křížová adsorbce:
    No
  • Formulář:
    Lyophilized
  • Synonyma antigenu:
    ATF|GDNF|Astrocyte-derived trophic factor|neurotrophic factor GDNF|Glial cell line-derived neurotrophic factor
  • Počet aminokyselin:
    142 - 158
  • Sekvence:
    FRYCSGSCDAAETTYDK
  • Skladovací teplota:
    –20 °C
  • Transportní teplota:
    Ice
  • Imunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human GDNF (142-158aa FRYCSGSCDAAETTYDK), different from the related mouse sequence by three amino acids and rat sequence by two amino acids.
  • Čištění:
    purified by immunogen affinity purification
  • Rozměry protilátky:
    100 µg
  • Bal.:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Glial cell line-derived neurotrophic factor(GDNF) detection. Tested with WB, IHC-P, ICC in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: GDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat