Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MEK1 Rabbit Polyclonal Antibody
Anti-MEK1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1376
Supplier:  Boster Bio
undefined
Anti-MEK1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1376
Supplier:  Boster Bio

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Mitogen-activated protein kinase Kinase 1
  • Symbol antigenu:
    MEK1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    C-Terminal
  • Křížová adsorbce:
    No
  • Formulář:
    Lyophilized
  • Počet aminokyselin:
    353 - 367
  • Sekvence:
    KQLMVHAFIKRSDAE
  • Skladovací teplota:
    –20 °C
  • Transportní teplota:
    Ice
  • Imunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human MEK1 (353-367aa KQLMVHAFIKRSDAE), identical to the related mouse and rat sequences.
  • Čištění:
    purified by immunogen affinity purification
  • Rozměry protilátky:
    100 µg
  • Bal.:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Dual specificity mitogen-activated protein kinase kinase 1(MAP2K1) detection. Tested with WB, IHC-P, ICC in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: MEK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat