Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MEK1 Mouse Monoclonal Antibody [clone: OTI4E1]
Anti-MEK1 Mouse Monoclonal Antibody [clone: OTI4E1]
Catalog # ABCAAB139343-100
Supplier:  Abcam
undefined
Anti-MEK1 Mouse Monoclonal Antibody [clone: OTI4E1]
Catalog # ABCAAB139343-100
Supplier:  Abcam

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Mitogen-activated protein kinase kinase 1
  • Symbol antigenu:
    MAP2K1
  • Klonovalita:
    Monoclonal
  • Klon:
    4E1
  • Spojení:
    Unconjugated
  • Hostitel:
    Mouse
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Sekvence:
    1286
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    100 µl
  • Typ balení:
    Vial
  • Bal.:
    100 µl

Specifications

About this item

Mouse monoclonal [OTI4E1] to MEK1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: Monoclonal
Clone: 40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: