Anti-pan Cytokeratin Mouse Monoclonal Antibody [clone: C-11] (APC (Allophycocyanin))
Catalog # ABCAAB106166-100
Supplier: Abcam
Specifications
- Druh protilátky:Primární
- Název antigenu:Pan Cytokeratin
- Klonovalita:Monoclonal
- Klon:C-11
- Spojení:APC (Allophycocyanin)
- Hostitel:Mouse
- Izotyp:IgG1
- Sekvence:1286
- Rozměry protilátky:100 µg
- Typ balení:Vial
- Bal.:100 µG
Specifications
About this item
APC Mouse monoclonal [C-11] to pan Cytokeratin.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: pan Cytokeratin
Clonality: Monoclonal
Clone: C-11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human