Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-TREM1 Mouse Monoclonal Antibody [clone: 6B1]
Catalog # ABCAAB90808-50
Supplier:  Abcam
Anti-TREM1 Mouse Monoclonal Antibody [clone: 6B1]
Catalog # ABCAAB90808-50
Supplier:  Abcam

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Triggering receptor expressed on myeloid cells 1
  • Symbol antigenu:
    TREM1
  • Klonovalita:
    Monoclonal
  • Klon:
    6B1
  • Spojení:
    Unconjugated
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Mouse
  • Izotyp:
    IgG1
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    50 µg
  • Typ balení:
    Vial
  • Bal.:
    50 µG

Specifications

About this item

Mouse monoclonal [6B1] to TREM1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TREM1
Clonality: Monoclonal
Clone: 6B1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human