Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Dopamine D2 Receptor Rabbit Polyclonal Antibody
Anti-Dopamine D2 Receptor Rabbit Polyclonal Antibody
Catalog # ABCAAB85367-100
Supplier:  Abcam
undefined
Anti-Dopamine D2 Receptor Rabbit Polyclonal Antibody
Catalog # ABCAAB85367-100
Supplier:  Abcam

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Dopamine receptor D2
  • Symbol antigenu:
    DRD2
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat
  • Western blot:
    Yes
  • Sekvence:
    1286
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    100 µg
  • Typ balení:
    Vial
  • Bal.:
    100 µG

Specifications

About this item

Rabbit polyclonal to Dopamine D2 Receptor.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Dopamine D2 Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Human