Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Hsp104 Rabbit Polyclonal Antibody
  ABCAAB69549-200
 :  Abcam
Anti-Hsp104 Rabbit Polyclonal Antibody
  ABCAAB69549-200
 :  Abcam

 

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    HSP104
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunoprecipitace:
    Yes
  • Izotyp:
    IgG
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    200 µl
  • Bal.:
    200 µl

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Hsp104
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: