- Druh protilátky:Primární
- Název antigenu:Rho GTPase activating Protein 35
- Symbol antigenu:ARHGAP35
- Klonovalita:Monoclonal
- Klon:2389D3a
- Spojení:Unconjugated
- Hostitel:Mouse
- Izotyp:IgG1
- Reaktivita:Human
- Western blot:Yes
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:100 µg
- Bal.:100 µG
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ARHGAP35 (Rho GTPase activating protein 35)
Clonality: monoclonal
Clone: 2389D3a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human