Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Ocular development associated gene Mouse Monoclonal Antibody [clone: GATA9A1D3]
Anti-Ocular development associated gene Mouse Monoclonal Antibody [clone: GATA9A1D3]
Katalogové číslo ABCAAB50816-100
Dodavatel:  Abcam
Anti-Ocular development associated gene Mouse Monoclonal Antibody [clone: GATA9A1D3]
Katalogové číslo ABCAAB50816-100
Dodavatel:  Abcam

Specifikace

  • Druh protilátky:
    Primární
  • Název antigenu:
    Ocular development associated gene
  • Klonovalita:
    Monoclonal
  • Klon:
    GATA9A1D3
  • Spojení:
    Unconjugated
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Mouse
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG2b
  • Reaktivita:
    Human,
    Mouse
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

Specifikace

O tomto produktu

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Ocular development associated gene
Clonality: monoclonal
Clone: GATA9A1D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse