Anti-C3/C3b Rabbit Polyclonal Antibody (Biotinylated)
Katalogové číslo ABCAAB48342-150
Dodavatel: Abcam
Specifikace
- Druh protilátky:Primární
- Název antigenu:complement component 3
- Symbol antigenu:C3
- Klonovalita:Polyclonal
- Spojení:Biotin
- Hostitel:Rabbit
- Imunoprecipitace:Yes
- Izotyp:IgG
- Reaktivita:Human
- Western blot:Yes
- Sekvence:1322
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:150 µg
- Typ balení:Vial
- Bal.:150 µG
Specifikace
O tomto produktu
Biotin Rabbit polyclonal to C3.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: C3/C3b
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human