Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Bombesin Receptor 3 Rabbit polyclonal antibody unconjugated
Anti-Bombesin Receptor 3 Rabbit polyclonal antibody unconjugated
Katalogové číslo ABCAAB48274-100
Dodavatel:  Abcam
Anti-Bombesin Receptor 3 Rabbit polyclonal antibody unconjugated
Katalogové číslo ABCAAB48274-100
Dodavatel:  Abcam

Specifikace

  • Druh protilátky:
    Primární
  • Název antigenu:
    Bombesin Receptor 3
  • Symbol antigenu:
    BRS3
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    100 µl
  • Bal.:
    100 µl

Specifikace

O tomto produktu

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Bombesin Receptor 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human