Anti-STAT1 Rabbit Polyclonal Antibody
Katalogové číslo ABCAAB47425-100
Dodavatel: Abcam
Specifikace
- Druh protilátky:Primární
- Název antigenu:Signal Transducer And Activator of Transcription 1
- Symbol antigenu:STAT1
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Izotyp:IgG
- Reaktivita:Human,Mouse
- Western blot:Yes
- Sekvence:1286
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:100 µg
- Typ balení:Vial
- Bal.:100 µG
Specifikace
O tomto produktu
Rabbit polyclonal to STAT1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: STAT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human