Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CD151 Mouse Monoclonal Antibody [clone: 11G5a]
  ABCAAB33315-100
 :  Abcam
Anti-CD151 Mouse Monoclonal Antibody [clone: 11G5a]
  ABCAAB33315-100
 :  Abcam

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    CD151 molecule (Raph blood group)
  • Symbol antigenu:
    CD151
  • Klonovalita:
    Monoclonal
  • Klon:
    11G5a
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Mouse
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Imunoprecipitace:
    Yes
  • Izotyp:
    IgG1
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD151 (CD151 molecule (Raph blood group))
Clonality: monoclonal
Clone: 11G5a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human