Order Entry
Czech Republic
ContactUsLinkComponent
Anti-TPX2 Mouse Monoclonal Antibody [clone: 18D5-1]
Anti-TPX2 Mouse Monoclonal Antibody [clone: 18D5-1]
Katalogové číslo ABCAAB32795-100
Dodavatel:  Abcam
Anti-TPX2 Mouse Monoclonal Antibody [clone: 18D5-1]
Katalogové číslo ABCAAB32795-100
Dodavatel:  Abcam

Specifikace

  • Druh protilátky:
    Primární
  • Název antigenu:
    TPX2, Microtubule Nucleation Factor
  • Symbol antigenu:
    TPX2
  • Klonovalita:
    Monoclonal
  • Klon:
    18D5-1
  • Spojení:
    Unconjugated
  • Hostitel:
    Mouse
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Imunoprecipitace:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

Specifikace

O tomto produktu

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TPX2 (TPX2, microtubule-associated)
Clonality: monoclonal
Clone: 18D5-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat