Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-BID Rabbit monoclonal antibody unconjugated [clone: Y8]
  ABCAAB32060-100
 :  Abcam
Anti-BID Rabbit monoclonal antibody unconjugated [clone: Y8]
  ABCAAB32060-100
 :  Abcam

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    BH3 interacting domain death agonist
  • Symbol antigenu:
    BID
  • Klonovalita:
    Monoclonal
  • Klon:
    Y8
  • Spojení:
    Unconjugated
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Imunoprecipitace:
    Yes
  • Izotyp:
    IgG
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    100 µl
  • Bal.:
    100 µl

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: BID (BH3 interacting domain death agonist)
Clonality: monoclonal
Clone: Y8
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: