Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Plexin D1 Goat Polyclonal Antibody
Anti-Plexin D1 Goat Polyclonal Antibody
  ABCAAB28762-100
 :  Abcam
undefined
Anti-Plexin D1 Goat Polyclonal Antibody
  ABCAAB28762-100
 :  Abcam

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Plexin D1
  • Symbol antigenu:
    PLXND1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Goat
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Imunoprecipitace:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Mouse
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    100 µg
  • Typ balení:
    Vial
  • Bal.:
    100 µG

 

 

Goat polyclonal to Plexin D1.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Plexin D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse