Anti-TFIIF Rabbit Polyclonal Antibody
Catalog # ABCAAB28179-100
Supplier: Abcam
Specifications
- Druh protilátky:Primární
- Název antigenu:General transcription factor 2F, polypeptide 1, 74kDa
- Symbol antigenu:GTF2F1
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Imunoprecipitace:Yes
- Izotyp:IgG
- Reaktivita:Human
- Western blot:Yes
- Sekvence:1286
- ChIP:Yes
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:100 µg
- Typ balení:Vial
- Bal.:100 µG
Specifications
About this item
Rabbit polyclonal to TFIIF.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TFIIF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human