Anti-Semaphorin 7a Rabbit Polyclonal Antibody
ABCAAB23578-100
: Abcam
- Druh protilátky:Primární
- Název antigenu:Semaphorin 7a
- Symbol antigenu:SEMA7a
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Imunoprecipitace:Yes
- Izotyp:IgG
- Reaktivita:Rat,Mouse
- Western blot:Yes
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:100 µg
- Bal.:100 µG
:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Semaphorin 7a
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat