Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-HIV2 Nef Rat Monoclonal Antibody [clone: 4B5]
  ABCAAB22626-50
 :  Abcam
Anti-HIV2 Nef Rat Monoclonal Antibody [clone: 4B5]
  ABCAAB22626-50
 :  Abcam

 

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    HIV2 Nef
  • Klonovalita:
    Monoclonal
  • Klon:
    4B5
  • Spojení:
    Unconjugated
  • Hostitel:
    Rat
  • Imunoprecipitace:
    Yes
  • Izotyp:
    IgG1
  • Western blot:
    Yes
  • Sekvence:
    1202
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    50 µl
  • Typ balení:
    Vial
  • Bal.:
    50 µl

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HIV2 Nef
Clonality: monoclonal
Clone: 4B5
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: