Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Dopamine beta Hydroxylase Sheep Polyclonal Antibody
  ABCAAB19353-100
 :  Abcam
Anti-Dopamine beta Hydroxylase Sheep Polyclonal Antibody
  ABCAAB19353-100
 :  Abcam

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Dopamine Beta Hydroxylase
  • Symbol antigenu:
    DBH
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Sheep
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    100 µl
  • Bal.:
    100 µl

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Dopamine beta Hydroxylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: