Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Hsp27 (phospho S86) Rabbit Polyclonal Antibody
Anti-Hsp27 (phospho S86) Rabbit Polyclonal Antibody
Katalogové číslo ABCAAB17938-50
Dodavatel:  Abcam
undefined
Anti-Hsp27 (phospho S86) Rabbit Polyclonal Antibody
Katalogové číslo ABCAAB17938-50
Dodavatel:  Abcam

Specifikace

  • Druh protilátky:
    Primární
  • Název antigenu:
    Heat shock 27kDa protein 1
  • Symbol antigenu:
    HSPB1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    50 µl
  • Bal.:
    50 µl

Specifikace

O tomto produktu

Rabbit polyclonal to Hsp27 (phospho S86).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Hsp27
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human