Specifications
- Druh protilátky:Primární
- Název antigenu:acyl-CoA dehydrogenase, C-4 to C-12 straight chain
- Symbol antigenu:ACADM
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Goat
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human,Mouse
- Western blot:Yes
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ACADM (acyl-CoA dehydrogenase, C-4 to C-12 straight chain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse